Learn More
Ethyl cis-2-amino-1-cyclopentane carboxylate hydrochloride, 99%, Thermo Scientific Chemicals
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 97.0°C to 100.0°C |
CAS | 142547-15-7 |
Color | White |
Infrared Spectrum | Authentic |
Assay Percent Range | 99% |
Molecular Formula | C8H16ClNO2 |
Packaging | Glass Bottle |
MDL Number | MFCD00800503 |
Synonym | ethyl cis-2-amino-1-cyclopentane carboxylate hydrochloride, ethyl 1r,2s-2-aminocyclopentane-1-carboxylate hydrochloride, cis-2-amino-cyclopentanecarboxylic acid ethyl ester hydrochloride, rel-1r,2s-ethyl 2-aminocyclopentanecarboxylate hydrochloride, 1r,2s-rel-ethyl 2-aminocyclopentanecarboxylate hydrochloride, ethyl cis-2-aminocyclopentanecarboxylate hcl, cis-ethyl 2-amino-1-cyclopentanecarboxylate hydrochloride, 1r,2s-rel-ethyl2-aminocyclopentanecarboxylatehydrochloride, cis-2-amino-cyclopentanecarboxylic acid ethyl ester hydrochlorid, 1r*,2s*-ethyl 2-aminocyclopentane-1-carboxylate hydrochloride |
Solubility Information | Solubility in water: soluble in water |
Show More |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0296050 | Certificate of Analysis | 02/05/2023 | 298392500 |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.