Learn More
Ethyl indole-2-carboxylate, 98+%, Thermo Scientific Chemicals
49.90 € - 163.00 €
Chemical Identifiers
CAS | 3770-50-1 |
---|---|
Molecular Formula | C11H11NO2 |
Molecular Weight (g/mol) | 189.21 |
MDL Number | MFCD00005609 |
InChI Key | QQXQAEWRSVZPJM-UHFFFAOYSA-N |
Synonym | ethyl indole-2-carboxylate, indole-2-carboxylic acid ethyl ester, 2-carbethoxyindole, 1h-indole-2-carboxylic acid, ethyl ester, indole-2-carboxylic acid, ethyl ester, 1h-indole-2-carboxylic acid ethyl ester, ethylindole-2-carboxylate, ethyl 2-indolecarboxylate, 1h-indole-2-carboxylic acid,ethyl ester, 2-carboethoxyindole |
PubChem CID | 73125 |
IUPAC Name | ethyl 1H-indole-2-carboxylate |
SMILES | CCOC(=O)C1=CC2=CC=CC=C2N1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11360224
|
Thermo Scientific Alfa Aesar
L00797.06 |
5 g |
49.90 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11370224
|
Thermo Scientific Alfa Aesar
L00797.14 |
25 g |
163.00 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
3770-50-1 | |
189.21 | |
QQXQAEWRSVZPJM-UHFFFAOYSA-N | |
73125 | |
CCOC(=O)C1=CC2=CC=CC=C2N1 |
C11H11NO2 | |
MFCD00005609 | |
ethyl indole-2-carboxylate, indole-2-carboxylic acid ethyl ester, 2-carbethoxyindole, 1h-indole-2-carboxylic acid, ethyl ester, indole-2-carboxylic acid, ethyl ester, 1h-indole-2-carboxylic acid ethyl ester, ethylindole-2-carboxylate, ethyl 2-indolecarboxylate, 1h-indole-2-carboxylic acid,ethyl ester, 2-carboethoxyindole | |
ethyl 1H-indole-2-carboxylate |
Specifications
3770-50-1 | |
C11H11NO2 | |
146395 | |
QQXQAEWRSVZPJM-UHFFFAOYSA-N | |
ethyl 1H-indole-2-carboxylate | |
73125 | |
≥98% |
122°C to 125°C | |
MFCD00005609 | |
ethyl indole-2-carboxylate, indole-2-carboxylic acid ethyl ester, 2-carbethoxyindole, 1h-indole-2-carboxylic acid, ethyl ester, indole-2-carboxylic acid, ethyl ester, 1h-indole-2-carboxylic acid ethyl ester, ethylindole-2-carboxylate, ethyl 2-indolecarboxylate, 1h-indole-2-carboxylic acid,ethyl ester, 2-carboethoxyindole | |
CCOC(=O)C1=CC2=CC=CC=C2N1 | |
189.21 | |
189.21 | |
Ethyl indole-2-carboxylate |
Safety and Handling
missing translation for 'einecsNumber' : 223-206-4
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.