missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl isobutyrylacetate, 94%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
Description
Ethyl Isobutyrylacetate is used in the synthesis of piperazine derivatives as possible multireceptor atypical antipsychotics, affecting dopamine and serotonin receptor properties . Also used in the synthesis of pyrazinecarboxamide DGAT1 (diacylglycerol acyltransferase 1) inhibitors used in the treatment of obesity.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Ethyl Isobutyrylacetate is used in the synthesis of piperazine derivatives as possible multireceptor atypical antipsychotics, affecting dopamine and serotonin receptor properties . Also used in the synthesis of pyrazinecarboxamide DGAT1 (diacylglycerol acyltransferase 1) inhibitors used in the treatment of obesity.
Solubility
Soluble in chloroform and methanol.
Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Ethyl Isobutyrylacetate is used in the synthesis of piperazine derivatives as possible multireceptor atypical antipsychotics, affecting dopamine and serotonin receptor properties . Also used in the synthesis of pyrazinecarboxamide DGAT1 (diacylglycerol acyltransferase 1) inhibitors used in the treatment of obesity.
Solubility
Soluble in chloroform and methanol.
Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications
Specifications
Melting Point | -9°C |
Density | 0.98 |
Boiling Point | 172°C to 174°C |
Flash Point | 53°C (127°F) |
Refractive Index | 1.4265 |
Quantity | 5 g |
UN Number | UN3272 |
Beilstein | 636726 |
Solubility Information | Soluble in chloroform and methanol. |
Formula Weight | 158.2 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'