missing translation for 'onlineSavingsMsg'
Learn More

Ethyl isobutyrylacetate, 94%, Thermo Scientific Chemicals

Product Code. 11342836
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Ethyl Isobutyrylacetate is used in the synthesis of piperazine derivatives as possible multireceptor atypical antipsychotics, affecting dopamine and serotonin receptor properties . Also used in the synthesis of pyrazinecarboxamide DGAT1 (diacylglycerol acyltransferase 1) inhibitors used in the treatment of obesity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl Isobutyrylacetate is used in the synthesis of piperazine derivatives as possible multireceptor atypical antipsychotics, affecting dopamine and serotonin receptor properties . Also used in the synthesis of pyrazinecarboxamide DGAT1 (diacylglycerol acyltransferase 1) inhibitors used in the treatment of obesity.

Solubility
Soluble in chloroform and methanol.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7152-15-0
Molecular Formula C8H14O3
Molecular Weight (g/mol) 158.197
MDL Number MFCD00009198
InChI Key XCLDSQRVMMXWMS-UHFFFAOYSA-N
Synonym ethyl isobutyrylacetate, 4-methyl-3-oxo-pentanoic acid ethyl ester, pentanoic acid, 4-methyl-3-oxo-, ethyl ester, ethyl isobutyroylacetate, valeric acid, 4-methyl-3-oxo-, ethyl ester, .gamma.,.gamma.-dimethylacetoacetic acid ethyl ester, beta-keto-gamma-methylvaleric acid ethyl ester, ethyl alpha-isobutyroylacetate, ethylisobutyrylacetate, ethyl iso-butyrylacetate
PubChem CID 81583
IUPAC Name ethyl 4-methyl-3-oxopentanoate
SMILES CCOC(=O)CC(=O)C(C)C

Specifications

Melting Point -9°C
Density 0.98
Boiling Point 172°C to 174°C
Flash Point 53°C (127°F)
Refractive Index 1.4265
Quantity 25 g
UN Number UN3272
Beilstein 636726
Solubility Information Soluble in chloroform and methanol.
Formula Weight 158.2
Percent Purity 94%
Chemical Name or Material Ethyl isobutyrylacetate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'