Learn More
Ethyl malonyl chloride, 90%, tech., Thermo Scientific Chemicals
56.53 € - 528.00 €
Chemical Identifiers
CAS | 36239-09-5 |
---|---|
Molecular Formula | C5H7ClO3 |
Molecular Weight (g/mol) | 150.56 |
MDL Number | MFCD00000736 |
InChI Key | KWFADUNOPOSMIJ-UHFFFAOYSA-N |
Synonym | ethyl 3-chloro-3-oxopropionate, ethyl chloroformylacetate, ethyl chloroformyl acetate, propanoic acid, 3-chloro-3-oxo-, ethyl ester, ethyl3-chloro-3-oxopropanoate, ethyl malonylchloride, carbethoxyacetyl chloride, 2-ethoxycarbonylacetyl chloride, ethyl 2-chlorocarbonyl acetate, chloroformyl acetic acid ethyl ester |
PubChem CID | 118931 |
IUPAC Name | ethyl 3-chloro-3-oxopropanoate |
SMILES | CCOC(=O)CC(Cl)=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10743561
|
Thermo Scientific Acros
172690050 |
5 g | Glass bottle | N/A | |||||
10553541
|
Thermo Scientific Acros
172690250 |
25 g | Glass bottle | N/A | |||||
11995031
|
Thermo Scientific Acros
172691000 |
100 g | Glass bottle | N/A | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
36239-09-5 | |
150.56 | |
KWFADUNOPOSMIJ-UHFFFAOYSA-N | |
118931 | |
CCOC(=O)CC(Cl)=O |
C5H7ClO3 | |
MFCD00000736 | |
ethyl 3-chloro-3-oxopropionate, ethyl chloroformylacetate, ethyl chloroformyl acetate, propanoic acid, 3-chloro-3-oxo-, ethyl ester, ethyl3-chloro-3-oxopropanoate, ethyl malonylchloride, carbethoxyacetyl chloride, 2-ethoxycarbonylacetyl chloride, ethyl 2-chlorocarbonyl acetate, chloroformyl acetic acid ethyl ester | |
ethyl 3-chloro-3-oxopropanoate |
Specifications
36239-09-5 | |
1.1700g/mL | |
67°C | |
90% | |
ClCOCH2CO2C2H5 | |
MFCD00000736 | |
1.17 | |
Solubility in water: reacts | |
CCOC(=O)CC(Cl)=O | |
150.56 | |
150.56 | |
Liquid |
Yellow to Brown | |
79.0°C to 80.0°C (25.0 mmHg) | |
Authentic | |
C5H7ClO3 | |
1.4250 to 1.4310 | |
02,582 | |
ethyl 3-chloro-3-oxopropionate, ethyl chloroformylacetate, ethyl chloroformyl acetate, propanoic acid, 3-chloro-3-oxo-, ethyl ester, ethyl3-chloro-3-oxopropanoate, ethyl malonylchloride, carbethoxyacetyl chloride, 2-ethoxycarbonylacetyl chloride, ethyl 2-chlorocarbonyl acetate, chloroformyl acetic acid ethyl ester | |
KWFADUNOPOSMIJ-UHFFFAOYSA-N | |
ethyl 3-chloro-3-oxopropanoate | |
118931 | |
90% | |
Ethyl malonyl chloride |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Reacts violently with water.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 252-934-5
- Ethyl malonyl chloride
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- EUH014-Reacts violently with water.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.