missing translation for 'onlineSavingsMsg'
Learn More
Ethyl methanesulfonate, 99%, Thermo Scientific Chemicals Product Code.: 11478930

Ethyl methanesulfonate, 99%, Thermo Scientific Chemicals

Product Code. 11478930
5 g
Click to view available options
Quantity:
5 g
10 g
50 g
250 g
This item is not returnable. View return policy

Product Code. 11478930

Brand: Thermo Scientific Alfa Aesar A12938.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Ethyl methanesulfonate is used as mutagen for both mammalian and plant cells. It finds application as a model alkylating agent in the study of deoxyribonucleic acid (DNA) repair processes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl methanesulfonate is used as mutagen for both mammalian and plant cells. It finds application as a model alkylating agent in the study of deoxyribonucleic acid (DNA) repair processes.

Solubility
Miscible with water.

Notes
Special handling precautions required. View MSDS prior to purchase. MSDS available online at www.alfa.com.Moisture sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62-50-0
Molecular Formula C3H8O3S
Molecular Weight (g/mol) 124.154
MDL Number MFCD00007559
InChI Key PLUBXMRUUVWRLT-UHFFFAOYSA-N
Synonym ethyl mesylate, ethyl methanesulphonate, half-myleran, methanesulfonic acid, ethyl ester, methanesulfonic acid ethyl ester, methylsulfonic acid, ethyl ester, rcra waste number u119, ethyl methansulphonate, ethyl ester of methylsulfonic acid, ethyl ester of methanesulfonic acidShow MoreShow Less
PubChem CID 6113
ChEBI CHEBI:23994
IUPAC Name ethyl methanesulfonate
SMILES CCOS(=O)(=O)C

Specifications

Density 1.17
Boiling Point 85°C to 86°C (10 mmHg)
Flash Point 100°C (212°F)
Assay Percent Range 99%
Odor Fruit-like
Refractive Index 1.418
Quantity 5 g
UN Number UN2810
Beilstein 773969
Sensitivity Moisture sensitive
Merck Index 14,3827
Solubility Information Miscible with water.
Formula Weight 124.16
Percent Purity 99%
Chemical Name or Material Ethyl methanesulfonate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10233916 Certificate TypeCertificate of Analysis Date30/08/2024 Catalog NumberA12938.30,A12938.09,A12938.06,A12938.18
Lot Number10241484 Certificate TypeCertificate of Analysis Date23/06/2022 Catalog NumberA12938.30,A12938.09,A12938.06,A12938.18
Lot Number10205164 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA12938.30,A12938.09,A12938.06,A12938.18
Lot Number10124120 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA12938.30,A12938.09,A12938.06,A12938.18
Lot Number10161448 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA12938.30,A12938.09,A12938.06,A12938.18
compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 1B
  • Germ cell mutagenicity Category 1B
  • Reproductive toxicity Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H340-May cause genetic defects.
  • H350-May cause cancer.
  • H361f-Suspected of damaging fertility.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ethyl methanesulfonate, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.