missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl methanesulfonate, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
25 g
100 g
500 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Specifications
Density | 1.1600g/mL |
Boiling Point | 213.0°C to 214.0°C |
Flash Point | 100°C |
Assay Percent Range | 99% |
Packaging | Glass bottle |
Linear Formula | CH3SO3C2H5 |
Refractive Index | 1.4170 to 1.4190 |
Quantity | 500 g |
Beilstein | 04,5 |
Merck Index | 15,3882 |
Show More |
Safety and Handling
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS07_Warning.png)
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS08_Dangerous_Health_Hazard.png)
Product Identifier
- Ethyl methanesulfonate
- Danger
- Acute toxicity Category 4
- Carcinogenicity Category 1A
- Germ cell mutagenicity Category 1A
- H340-May cause genetic defects.
- H350-May cause cancer.
- H302-Harmful if swallowed.
- P201-Obtain special instructions before use.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'