missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl Methyl Ketone, Extra Pure, SLR, Fisher Chemical™
Click to view available options
Quantity:
500 mL
1 L
2.5 L
5 L
25 L
Packaging:
Amber glass bottle
Metal can
Plastic Coated Glass bottle
Description
C4H8O, CAS Number-78-93-3, methyl ethyl ketone, methylethylketone, methyl acetone, 2-butanone, methylethyl ketone, meetco, butanone, 3-butanone, ethyl methyl ketone, ethylmethylketon, 2.5L, 80 deg.C, CHEBI:28398, Colorless, 72.11g/mol, ZWEHNKRNPOVVGH-UHFFFAOYSA-N, butan-2-one, -87 deg.C, 11648
Specifications
Specifications
Melting Point | -87°C |
Color | Colorless |
Boiling Point | 80°C |
Packaging | Amber glass bottle |
Quantity | 1 L |
Formula Weight | 72.11g/mol |
Physical Form | Liquid |
Safety and Handling
Product Identifier
- 2-Butanone
- Danger
- Flammable liquid Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
- EUH066-Repeated exposure may cause skin dryness or cracking.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P240-Ground/bond container and receiving equipment.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction