missing translation for 'onlineSavingsMsg'
Learn More

Ethyl N-(2,4-dinitrophenoxy)acetimidate, 98+%, Thermo Scientific™

Product Code. 11401240
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54322-32-6
Molecular Formula C10H11N3O6
Molecular Weight (g/mol) 269.213
MDL Number MFCD00010133
InChI Key UVBZLMGBNXCYNA-YRNVUSSQSA-N
Synonym ethyl n-2,4-dinitrophenoxy acetimidate, pubchem12897, jlyppahrtackn bt jw dtttrrvbapemaadmu@dmp, ethyl e-n-2,4-dinitrophenoxy acetimidate, 1-1e-2-ethoxy-1-azaprop-1-enyloxy-2,4-dinitrobenzene
PubChem CID 9566065
IUPAC Name ethyl (1E)-N-(2,4-dinitrophenoxy)ethanimidate
SMILES CCOC(=NOC1=C(C=C(C=C1)[N+](=O)[O-])[N+](=O)[O-])C

Specifications

Melting Point 110°C to 113°C
Quantity 5 g
Sensitivity Moisture sensitive
Formula Weight 269.21
Percent Purity ≥98%
Chemical Name or Material Ethyl N-(2,4-dinitrophenoxy)acetimidate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'