missing translation for 'onlineSavingsMsg'
Learn More

Ethyl oxalyl chloride, 98%

Product Code. 10254960 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Packaging:
Glass bottle
Unit Size:
100mL
25mL
500mL
This item is not returnable. View return policy

Product Code. 10254960

Brand: Thermo Scientific Acros 118781000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4755-77-5 | C4H5ClO3 | 136.53 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4755-77-5
Molecular Formula C4H5ClO3
Molecular Weight (g/mol) 136.53
MDL Number MFCD00000706
InChI Key OWZFULPEVHKEKS-UHFFFAOYSA-N
Synonym ethyl chlorooxoacetate, ethyl oxalyl chloride, ethyl chloroglyoxylate, ethoxalyl chloride, ethyl oxalyl monochloride, acetic acid, chlorooxo-, ethyl ester, oxalic acid monoethyl ester chloride, monoethyl oxaloyl chloride, oxalyl chloride, ethyl ester, ethyl chlorocarbonyl formate
PubChem CID 20884
IUPAC Name ethyl 2-chloro-2-oxoacetate
SMILES CCOC(=O)C(=O)Cl

Specifications

Density 1.2200g/mL
Boiling Point 135°C
Flash Point 41°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula ClCOCO2C2H5
Refractive Index 1.4160 to 1.418
Quantity 100 mL
Beilstein 02, 541
Specific Gravity 1.22
Solubility Information Solubility in water: decomposes. Other solubilities: soluble in common organic solvents
Formula Weight 136.53
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Ethyl oxalyl chloride
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.