missing translation for 'onlineSavingsMsg'
Learn More

Ethyl pentafluoropropionate, 98+%, Thermo Scientific Chemicals

Product Code. 11361407
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 426-65-3
Molecular Formula C5H5F5O2
Molecular Weight (g/mol) 192.085
MDL Number MFCD00000431
InChI Key DBOFMRQAMAZKQY-UHFFFAOYSA-N
Synonym ethyl pentafluoropropionate, ethyl perfluoropropionate, ethylpentafluoropropionate, ethyl pentafluoropropanoate, pentafluoropropionic acid ethyl ester, propanoic acid, pentafluoro-, ethyl ester, propanoic acid, 2,2,3,3,3-pentafluoro-, ethyl ester, 2,2,3,3,3-pentafluoro-propionic acid ethyl ester, pubchem12575, acmc-1asrx
PubChem CID 67928
IUPAC Name ethyl 2,2,3,3,3-pentafluoropropanoate
SMILES CCOC(=O)C(C(F)(F)F)(F)F

Specifications

Density 1.299
Boiling Point 73°C to 74°C
Flash Point 1°C (33°F)
Refractive Index 1.301
Quantity 25 g
UN Number UN3272
Beilstein 1779789
Formula Weight 192.09
Percent Purity ≥98%
Chemical Name or Material Ethyl pentafluoropropionate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'