missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl perfluorooctanoate, 97%, Thermo Scientific™
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
5 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Specifications
Density | 1.626 |
Boiling Point | 167°C to 168°C |
Flash Point | None |
Linear Formula | CF3(CF2)6CO2CH2CH3 |
Refractive Index | 1.311 |
Quantity | 5 g |
Beilstein | 1810008 |
Synonym | ethyl perfluorooctanoate, ethyl perfluorocaprylate, octanoic acid, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecafluoro-, ethyl ester, octanoic acid,2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecafluoro-, ethyl ester, acmc-20akwv, ethyl perfluorooctanoat, ethyl perfluorooctanonate, octanoic acid, pentadecafluoro-, ethyl ester, pentadecafluorooctanoic acid ethyl ester, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecafluoro-octanoic acid ethyl ester |
Formula Weight | 442.12 |
Percent Purity | 97% |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'