missing translation for 'onlineSavingsMsg'
Learn More

Ethyl phenylacetate, 99%, Thermo Scientific Chemicals

Product Code. 10356790
Click to view available options
Quantity:
5 g
250 g
1 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 101-97-3
Molecular Formula C10H12O2
Molecular Weight (g/mol) 164.20
InChI Key DULCUDSUACXJJC-UHFFFAOYSA-N
IUPAC Name ethyl 2-phenylacetate
SMILES CCOC(=O)CC1=CC=CC=C1

Specifications

Melting Point -29.0°C
Density 1.0300 g/mL
Boiling Point 226.0°C
Flash Point 102°C
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula C6H5CH2CO2C2H5
Refractive Index 1.4970 to 1.4990
Quantity 1 kg
Beilstein 09, 434
Merck Index 15, 3893
Specific Gravity 1.03
Solubility Information Solubility aq. soln.: insoluble. Other solubilities: very soluble in ethanol and diethyl ether, soluble in chloroform
Formula Weight 164.2
Chemical Name or Material Ethyl phenylacetate
Show More Show Less
Product Identifier
  • Ethyl phenylacetate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'