missing translation for 'onlineSavingsMsg'
Learn More

Ethyl propionylacetate, 98%, Thermo Scientific Chemicals

Product Code. 10751701
Click to view available options
Quantity:
1 mL
5 mL
25 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4949-44-4
Molecular Formula C7H12O3
Molecular Weight (g/mol) 144.17
MDL Number MFCD00009317
InChI Key UDRCONFHWYGWFI-UHFFFAOYSA-N
Synonym ethyl propionylacetate, ethyl 3-oxovalerate, ethyl 3-oxo-n-valerate, pentanoic acid, 3-oxo-, ethyl ester, 3-oxopentanoic acid ethyl ester, 3-oxo-pentanoic acid ethyl ester, ethyl 3-ketopentanoate, 3-oxovaleric acid ethyl ester, propionylacetic acid ethyl ester, ethylpropanylacetate
PubChem CID 78656
IUPAC Name ethyl 3-oxopentanoate
SMILES CCC(=O)CC(=O)OCC

Specifications

Color Colorless to Yellow
Density 1.0100g/mL
Boiling Point 83.0°C to 84.0°C (12.0 mmHg)
Flash Point 77°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Linear Formula C2H5COCH2CO2C2H5
Refractive Index 1.4220 to 1.4240
Quantity 5 mL
Specific Gravity 1.01
Formula Weight 144.17
Percent Purity 98+%
Physical Form Liquid
Chemical Name or Material Ethyl propionylacetate
Show More Show Less
compliance-icons
Product Identifier
  • Ethyl propionylacetate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'