missing translation for 'onlineSavingsMsg'
Learn More
Ethyl pyrazolo[1,5-a]pyridine-3-carboxylate, 95%, Thermo Scientific™ Product Code.: 10709163

Ethyl pyrazolo[1,5-a]pyridine-3-carboxylate, 95%, Thermo Scientific™

Product Code. 10709163
1 g, Amber Glass Bottle
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy

Product Code. 10709163

Brand: Thermo Scientific Maybridge CC26523DA

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 16205-44-0
Molecular Formula C10H10N2O2
Molecular Weight (g/mol) 190.20
MDL Number MFCD06411542
InChI Key IDHMFSBXNNIWCT-UHFFFAOYSA-N
Synonym ethyl pyrazolo 1,5-a pyridine-3-carboxylate, pyrazolo 1,5-a pyridine-3-carboxylic acid ethyl ester, ethylpyrazolo 1,5-a pyridine-3-carboxylate, ethyl h-pyrazolo 1,5-a pyridine-3-carboxylate, zlchem 54, acmc-1bsao, ethyl 3-azaindoline-3-carboxylate, ethyl 8-hydropyrazolo 1,5-a pyridine-3-carboxylate, 3-pyrazolo 1,5-a pyridinecarboxylic acid ethyl ester, 1qp
PubChem CID 2795464
IUPAC Name ethyl pyrazolo[1,5-a]pyridine-3-carboxylate
SMILES CCOC(=O)C1=C2C=CC=CN2N=C1

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 190.2
Percent Purity 95%
Chemical Name or Material Ethyl pyrazolo[1, 5-a]pyridine-3-carboxylate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ethyl pyrazolo[1,5-a]pyridine-3-carboxylate, 95%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.