missing translation for 'onlineSavingsMsg'
Learn More

Ethylboronic acid, 95%, Thermo Scientific Chemicals

49.65 € - 155.78 €

Chemical Identifiers

CAS 4433-63-0
Molecular Formula C2H7BO2
Molecular Weight (g/mol) 73.89
MDL Number MFCD01074536
InChI Key PAVZHTXVORCEHP-UHFFFAOYSA-N
Synonym boronic acid, ethyl, ethaneboronic acid, ethyldihydroxyborane, ethyl boronic acid, boronic acid, ethyl-9ci, ethylboronicacid, pubchem7960, ethylboric acid, etb oh 2
PubChem CID 521157
IUPAC Name ethylboronic acid
SMILES CCB(O)O
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10772643
View Documents
Thermo Scientific Acros
370860010
1 g Glass bottle
49.65 €
1g
Please to purchase this item. Need a web account? Register with us today!
11965851
View Documents
Thermo Scientific Acros
370860050
5 g Glass bottle
155.78 €
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

4433-63-0
73.89
PAVZHTXVORCEHP-UHFFFAOYSA-N
521157
CCB(O)O
C2H7BO2
MFCD01074536
boronic acid, ethyl, ethaneboronic acid, ethyldihydroxyborane, ethyl boronic acid, boronic acid, ethyl-9ci, ethylboronicacid, pubchem7960, ethylboric acid, etb oh 2
ethylboronic acid

Specifications

4433-63-0
Authentic
C2H7BO2
MFCD01074536
PAVZHTXVORCEHP-UHFFFAOYSA-N
ethylboronic acid
521157
95%
Ethylboronic acid
White to Yellow
95%
CH3CH2B(OH)2
boronic acid, ethyl, ethaneboronic acid, ethyldihydroxyborane, ethyl boronic acid, boronic acid, ethyl-9ci, ethylboronicacid, pubchem7960, ethylboric acid, etb oh 2
CCB(O)O
73.89
73.89
Crystalline Powder, Flakes or Needles
Videos
Safety and Handling

Safety and Handling

GHS Signal Word: Warning

Product Identifier
  • Ethylboronic acid

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.