missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ethyle2-({[(2-ethoxy-2-oxoethyl)thio]methyl}thio)acetate, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
10 g
50 g
Specifications
Specifications
CAS Min % | 97.0 |
Assay Percent Range | 97% |
Packaging | Amber glass bottle |
Linear Formula | CH3CH2O2CCH2SCH2SCH2CO2CH2CH3 |
Quantity | 10 g |
Formula Weight | 252.35 |
Percent Purity | 97% |
Chemical Name or Material | Ethyl 2-({[(2-ethoxy-2-oxoethyl)thio]methyl}thio)acetate |
Safety and Handling
Product Identifier
- Ethyl 2-({¢(2-ethoxy-2-oxoethyl)thio!methyl}thio)acetate
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'