missing translation for 'onlineSavingsMsg'
Learn More

ethyle2-chloro-3,4-dimethoxybenzoate, 97%, Thermo Scientific™

Product Code. 10475492
Click to view available options
Quantity:
10 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Specifications

CAS 175136-04-6
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C11H13ClO4
MDL Number MFCD00085062
Quantity 10 g
Synonym maybridge1_002113
InChI Key AXNMDQHZRJDVRG-UHFFFAOYSA-N
SMILES CCOC(=O)C1=C(Cl)C(OC)=C(OC)C=C1
IUPAC Name ethyl 2-chloro-3,4-dimethoxybenzoate
Molecular Weight (g/mol) 244.67
PubChem CID 2774987
Formula Weight 244.67
Percent Purity 97%
Chemical Name or Material Ethyl 2-chloro-3, 4-dimethoxybenzoate
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'