missing translation for 'onlineSavingsMsg'
Learn More
ethyle4-formyl-2,5-dimethyl-1-phenyl-1H-pyrrole-3-carboxylate, 97%, Thermo Scientific™ Product Code.: 10618925

ethyle4-formyl-2,5-dimethyl-1-phenyl-1H-pyrrole-3-carboxylate, 97%, Thermo Scientific™

Product Code. 10618925
5 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 10618925

Brand: Thermo Scientific Maybridge SP00028DE

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 175276-52-5
Molecular Formula C16H17NO3
Molecular Weight (g/mol) 271.316
MDL Number MFCD00204225
InChI Key BGDYIZGTPVRWAM-UHFFFAOYSA-N
Synonym ethyl 4-formyl-2,5-dimethyl-1-phenyl-1h-pyrrole-3-carboxylate, 4-formyl-2,5-dimethyl-1-phenyl-1h-pyrrole-3-carboxylic acid ethyl esterShow MoreShow Less
PubChem CID 2798484
IUPAC Name ethyl 4-formyl-2,5-dimethyl-1-phenylpyrrole-3-carboxylate
SMILES CCOC(=O)C1=C(N(C(=C1C=O)C)C2=CC=CC=C2)C

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Quantity 5 g
Formula Weight 271.31
Percent Purity 97%
Chemical Name or Material Ethyl 4-formyl-2, 5-dimethyl-1-phenyl-1H-pyrrole-3-carboxylate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
ethyle4-formyl-2,5-dimethyl-1-phenyl-1H-pyrrole-3-carboxylate, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.