missing translation for 'onlineSavingsMsg'
Learn More

ethyle5-(2-bromoacetyl)isoxazole-3-carboxylate, 97%, Thermo Scientific™

Product Code. 10052264
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 104776-74-1
Molecular Formula C8H8BrNO4
Molecular Weight (g/mol) 262.06
MDL Number MFCD00173773
InChI Key GUYZRQOTADGHII-UHFFFAOYSA-N
Synonym ethyl 5-2-bromoacetyl isoxazole-3-carboxylate, ethyl 5-bromoacetyl isoxazole-3-carboxylate, ethyl 5-2-bromoacetyl-1,2-oxazole-3-carboxylate, ethyl 5-bromoacetyl-1,2-oxazole-3-carboxylate, 3-isoxazolecarboxylicacid, 5-2-bromoacetyl-, ethyl ester, acmc-20a3va, 5-bromoacetyl-3-ethoxycarbonyl isoxazole, ethyl 5-2-bromanylethanoyl-1,2-oxazole-3-carboxylate, 5-2-bromo-1-oxoethyl-3-isoxazolecarboxylic acid ethyl ester
PubChem CID 2736373
IUPAC Name ethyl 5-(2-bromoacetyl)-1,2-oxazole-3-carboxylate
SMILES CCOC(=O)C1=NOC(=C1)C(=O)CBr

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 262.06
Percent Purity 97%
Chemical Name or Material ethyle5-(2-bromoacetyl)isoxazole-3-carboxylate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'