missing translation for 'onlineSavingsMsg'
Learn More
ethyle5-(4-chlorophenyl)-2-formyl-3-furoate, 97%, Thermo Scientific™ Product Code.: 10792323

ethyle5-(4-chlorophenyl)-2-formyl-3-furoate, 97%, Thermo Scientific™

Product Code. 10792323
1 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 10792323

Brand: Thermo Scientific Maybridge SP00067DA

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 175276-64-9
Molecular Formula C14H11ClO4
Molecular Weight (g/mol) 278.688
MDL Number MFCD00275550
InChI Key XFOQMTOCKSVCBO-UHFFFAOYSA-N
PubChem CID 2798516
IUPAC Name ethyl 5-(4-chlorophenyl)-2-formylfuran-3-carboxylate
SMILES CCOC(=O)C1=C(OC(=C1)C2=CC=C(C=C2)Cl)C=O

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1 g
Synonym ethyl 5-4-chlorophenyl-2-formyl-3-furoate, ethyl 5-4-chlorophenyl-2-formylfuran-3-carboxylate, ethyl5-4-chlorophenyl-2-formyl-3-furoate, 3-furancarboxylic acid,5-4-chlorophenyl-2-formyl-, ethyl ester, 3-furancarboxylic acid,5-4-chlorophenyl-2-formyl-,ethyl ester
Formula Weight 278.69
Percent Purity 97%
Chemical Name or Material ethyle5-(4-chlorophenyl)-2-formyl-3-furoate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
ethyle5-(4-chlorophenyl)-2-formyl-3-furoate, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.