missing translation for 'onlineSavingsMsg'
Learn More

ethyle5-amino-1-(4-fluorophenyl)-1H-pyrazole-4-carboxylate, 97%, Thermo Scientific™

Product Code. 10031144
Click to view available options
Quantity:
1 g
10 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 138907-68-3
Molecular Formula C12H12FN3O2
Molecular Weight (g/mol) 249.25
MDL Number MFCD00173917
InChI Key RPPPCKSHIYWAPW-UHFFFAOYSA-N
Synonym ethyl 5-amino-1-4-fluorophenyl-1h-pyrazole-4-carboxylate, ethyl 5-amino-1-4-fluorophenyl pyrazole-4-carboxylate, ethyl 5-amino-1-4-fluoro-phenyl-1h-pyrazole-4-carboxylate, 5-amino-1-4-fluorophenyl-1h-pyrazole-4-carboxylic acid ethyl ester, 5-amino-4-ethoxycarbonyl-1-4-fluorophenyl-1h-pyrazole, 1-5-amino-4-ethoxycarbonyl-1h-pyrazol-1-yl-4-fluorobenzene, maybridge1_006170, ethyl5-amino-1-4-fluorophenyl-1h-pyrazole-4-carboxylate, ethylaminofluorophenylpyrazolecarboxylate, 5-amino-1-4-fluorophenyl-1h-pyrazole-4-carboxyli
PubChem CID 688689
IUPAC Name ethyl 5-amino-1-(4-fluorophenyl)pyrazole-4-carboxylate
SMILES CCOC(=O)C1=C(N)N(N=C1)C1=CC=C(F)C=C1

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Quantity 25 g
Formula Weight 249.24
Percent Purity 97%
Chemical Name or Material ethyle5-amino-1-(4-fluorophenyl)-1H-pyrazole-4-carboxylate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.