missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethylene carbonate, 99+%, Thermo Scientific Chemicals
missing translation for 'orderingAttributeHoverText'
Quantity:
250 g
1 kg
5 kg
Packaging:
Glass bottle
Plastic drum
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
Alkylation reagent
Specifications
Specifications
Melting Point | 34.0°C to 37.0°C |
Color | White to Yellow |
Density | 1.3210g/mL |
Boiling Point | 246.7°C |
Flash Point | 150°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 99% min. (GC) |
Packaging | Glass bottle |
Quantity | 1 kg |
Beilstein | 19, 100 |
Show More |
Safety and Handling
Product Identifier
- Ethylene carbonate
- Warning
- Serious eye damage/eye irritation Category 2
- H319-Causes serious eye irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'