missing translation for 'onlineSavingsMsg'
Learn More

Ethylene carbonate, 99%, Thermo Scientific Chemicals

Product Code. 11412378
Click to view available options
Quantity:
10,000 g
2500 g
500 g

Solvent for many polymers and resins, plasticisers, rubbers and textiles.Ethylene carbonate is used as a polar solvent and a high permittivity component of electrolytes in lithium batteries. It finds application in surface coatings, dyes, fibers and plastics. It is also used as a plasticizer and as a precursor to vinylene carbonate. Further, it is used in the preparation of dimethyl carbonate, which acts as a useful solvent and a mild methylating agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Solvent for many polymers and resins, plasticisers, rubbers and textiles.Ethylene carbonate is used as a polar solvent and a high permittivity component of electrolytes in lithium batteries. It finds application in surface coatings, dyes, fibers and plastics. It is also used as a plasticizer and as a precursor to vinylene carbonate. Further, it is used in the preparation of dimethyl carbonate, which acts as a useful solvent and a mild methylating agent.

Solubility
Miscible (40%) with water, alcohol ethyl acetate, benzene and chloroform. Soluble in ether, n-Butanol, and carbon tetrachloride.Soluble in water, n-butanol, ethyl acetate, benzene, carbon tetrachloride, ether and chloroform. Insoluble in gasoline and turpentine oil.

Notes
Incompatible with strong oxidizing agents, acids, bases and reducing agents. Store in a cool place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 96-49-1
Molecular Formula C3H4O3
Molecular Weight (g/mol) 88.062
MDL Number MFCD00005382
InChI Key KMTRUDSVKNLOMY-UHFFFAOYSA-N
Synonym ethylene carbonate, glycol carbonate, ethylene glycol carbonate, dioxolone-2, cyclic ethylene carbonate, ethylene carbonic acid, 1,3-dioxacyclopentan-2-one, 2-dioxolone, carbonic acid, cyclic ethylene ester, texacar ec
PubChem CID 7303
IUPAC Name 1,3-dioxolan-2-one
SMILES C1COC(=O)O1

Specifications

Melting Point 35°C to 37°C
Density 1.32
Boiling Point 244°C to 245°C
Flash Point 160°C (320°F)
Quantity 500 g
Beilstein 106249
Solubility Information Miscible (40%) with water,alcohol ethyl acetate,benzene and chloroform; Soluble in ether,n-Butanol,and carbon tetrachloride;Soluble in water,n-butanol,ethyl acetate,benzene,carbon tetrachloride,ether and chloroform; Insoluble in gasoline and turpentine oil.
Formula Weight 88.06
Percent Purity 99%
Chemical Name or Material Ethylene carbonate
compliance-icons
Product Identifier
  • Ethylene carbonate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : 1,3-Dioxolan-2-one

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.