missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethylene glycol, 99.5%, for analysis, Thermo Scientific Chemicals
76.34 EUR valid until 2024-08-31
Use promo code "21743" to get your promotional price.
Use promo code "21743" to get your promotional price.
Click to view available options
Quantity:
1 L
2.5 L
25 L
Packaging:
Glass Bottle
Metal Drum
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For analysis
Specifications
Melting Point | -13.0°C |
Density | 1.1130g/mL |
Boiling Point | 196.0°C to 198.0°C |
Quantity | 1 L |
Flash Point | 116°C |
Infrared Spectrum | Authentic |
Packaging | Glass Bottle |
Physical Form | Viscous Liquid |
Refractive Index | 1.4300 to 1.4330 |
Assay Percent Range | 99.5% |
Show More |
Safety and Handling
Product Identifier
- Ethylene glycol
- Warning
- Acute toxicity Category 4
- Specific target organ toxicity after repeated exposure Category 2
- H373-May cause damage to organs.
- H302-Harmful if swallowed.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'