missing translation for 'onlineSavingsMsg'
Learn More

Ethylene glycol, 99.8%, anhydrous, AcroSeal™, Thermo Scientific Chemicals

Product Code. 11934631
Click to view available options
Quantity:
100 mL
1 L

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107-21-1
Molecular Formula C2H6O2
Molecular Weight (g/mol) 62.06
InChI Key LYCAIKOWRPUZTN-UHFFFAOYSA-N
Synonym ethylene glycol, 1,2-ethanediol, glycol, monoethylene glycol, 1,2-dihydroxyethane, 2-hydroxyethanol, glycol alcohol, ethylene alcohol, fridex, tescol
PubChem CID 174
ChEBI CHEBI:30742
IUPAC Name ethane-1,2-diol
SMILES C(CO)O

Specifications

Melting Point -13.0°C
Density 1.1130g/mL
Boiling Point 196.0°C to 198.0°C
Flash Point 116°C
Infrared Spectrum Authentic
Assay Percent Range 99.75% min. (GC)
Packaging AcroSeal™ Glass bottle
Linear Formula HOCH2CH2OH
Refractive Index 1.4300 to 1.4330
Quantity 100 mL
Merck Index 15,3852
Specific Gravity 1.113
Solubility Information Solubility in water: miscible. Other solubilities: miscible with acetic acid, acetone, glycerol,, pyridine, lower aliphatic alcohols,, aldehydes and ketones, slightly soluble in ether, practically insoluble in benzene and hydrocarbons
Viscosity 21 mPa.s (20°C)
Formula Weight 62.06
Percent Purity 99.8%
Chemical Name or Material Ethylene glycol
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Ethylene glycol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H373-May cause damage to organs.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.