missing translation for 'onlineSavingsMsg'
Learn More

Ethylene glycol, 99+%, extra pure, Thermo Scientific Chemicals

Product Code. 10695572
Click to view available options
Quantity:
1 L
2.5 L
25 L
Packaging:
Glass Bottle
Plastic Drum

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107-21-1
Molecular Formula C2H6O2
Molecular Weight (g/mol) 62.068
MDL Number MFCD00002885
InChI Key LYCAIKOWRPUZTN-UHFFFAOYSA-N
Synonym ethylene glycol, 1,2-ethanediol, glycol, monoethylene glycol, 1,2-dihydroxyethane, 2-hydroxyethanol, glycol alcohol, ethylene alcohol, fridex, tescol
PubChem CID 174
ChEBI CHEBI:30742
IUPAC Name ethane-1,2-diol
SMILES C(CO)O

Specifications

Melting Point 2°C to 3°C
Decomposition Information >500°C
Density 1.1130g/mL
Boiling Point 196.0°C to 198.0°C
Quantity 2.5 L
Flash Point 116°C (240.8°F)
Infrared Spectrum Authentic
Packaging Glass Bottle
Physical Form Viscous Liquid
Refractive Index 1.4300 to 1.4330 (20°C, 589nm)
Assay Percent Range 99+%
Linear Formula HOCH2CH2OH
Vapor Pressure Vapor Pressure: 0.12mmHg at 20°C
Beilstein 01, 465
Water 0.1% max. (Coulometric)
Fieser 01,375; 05,296; 09,217; 15,156
Merck Index 15, 3852
Formula Weight 62.06
Percent Purity ≥99.0%
Grade Extra Pure
Chemical Name or Material Ethylene glycol, 99+%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Ethylene glycol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H373-May cause damage to organs.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.