missing translation for 'onlineSavingsMsg'
Learn More

Ethylene Glycol, BAKER ANALYZED™ Reagent, J.T. Baker™

Product Code. 15508574
missing translation for 'orderingAttributeHoverText'
Quantity:
500 mL
4 L
Packaging:
Narrow Mouth Amber Glass Bottle
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

High quality chemicals for laboratory and specialized industrial use

    For Laboratory,Research,or Manufacturing Use

Identifiants chimiques

CAS 107-21-1
Molecular Formula C2H6O2
Molecular Weight (g/mol) 62.068
InChI Key LYCAIKOWRPUZTN-UHFFFAOYSA-N
Synonym ethylene glycol, 1,2-ethanediol, glycol, monoethylene glycol, 1,2-dihydroxyethane, 2-hydroxyethanol, glycol alcohol, ethylene alcohol, fridex, tescol
PubChem CID 174
ChEBI CHEBI:30742
IUPAC Name ethane-1,2-diol
SMILES C(CO)O

Spécification

Quantity 4 L
Flash Point 111.0°C (231.8°F)
Packaging Narrow Mouth Amber Glass Bottle
Specific Gravity 1L = 1.10kg
Formula Weight 62.07
Grade BAKER ANALYZED™ Reagent
Chemical Name or Material Ethylene Glycol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'