missing translation for 'onlineSavingsMsg'
Learn More

Ethylene glycol dimethyl ether, 99.5%, Thermo Scientific Chemicals

58.79 € - 308.54 €

Specifications

Chemical Name or Material Ethylene glycol dimethyl ether
Melting Point -69.0°C
CAS 110-71-4
Density 0.8670g/mL
Boiling Point 84.0°C to 86.0°C
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11904731
View Documents
Thermo Scientific Acros
442981000
100 mL
58.79 €
100mL
Please to purchase this item. Need a web account? Register with us today!
11994721
View Documents
Thermo Scientific Acros
442980010
1 L
308.54 €
1 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Ethylene glycol dimethyl ether
110-71-4
84.0°C to 86.0°C
1.3785 to 1.3805
CH3OCH2CH2OCH3
0.005% max.
Solubility in water: miscible. Other solubilities: solulbe in hydrocarbon solvents, soluble in ethanol, ethyl ether, acetone, carbon, tetrachloride, benzene, dichloroethane
99.5%
-69.0°C
0.8670g/mL
−6°C
0.867
1.1 mPa.s (20°C)
monoglyme, ethylene glycol dimethyl ether, glyme, dimethyl cellosolve, egdme, ethane, 1,2-dimethoxy, dimethoxyethane, 2,5-dioxahexane, glycol dimethyl ether, ethylene dimethyl ether
90.12
Extra Dry over Molecular Sieve
Videos
Safety and Handling

Safety and Handling

GHS H Statement Harmful if inhaled. May damage fertility. May damage the unborn child. Highly flammable liquid and vapor. Causes skin irritation. May form explosive peroxides.

GHS P Statement Obtain special instructions before use. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician if you feel unwell. Wear protective gloves/protective clothing. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. IF ON SKIN: Wash with plenty of soap and water.

Danger

Product Identifier
  • Ethylene glycol dimethyl ether

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Flammable liquid Category 2
  • Reproductive toxicity Category 1B
  • Skin corrosion/irritation Category 2

Hazard Statement
  • H360FD-May damage fertility. May damage the unborn child.
  • H315-Causes skin irritation.
  • H332-Harmful if inhaled.
  • H225-Highly flammable liquid and vapour.
  • EUH019-May form explosive peroxides.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.