missing translation for 'onlineSavingsMsg'
Learn More

Ethylene glycol dimethyl ether, 99%, Extra Dry, AcroSeal(R), Thermo Scientific Chemicals

85.73 € - 298.49 €

Specifications

Chemical Name or Material Ethylene glycol dimethyl ether
CAS 128-37-0
Density 0.8670g/mL
Identification Pass Test
Packaging AcroSeal™ Glass Bottle
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10336744
View Documents
Thermo Scientific Acros
610351000
100 mL AcroSeal™ Glass Bottle
85.73 €
100mL
Please to purchase this item. Need a web account? Register with us today!
10614375
View Documents
Thermo Scientific Acros
610350010
1 L AcroSeal™ Glass Bottle
298.49 €
1 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Ethylene glycol dimethyl ether
0.8670g/mL
AcroSeal™ Glass Bottle
CH3OCH2CH2OCH3
01, 467
01,267; 02,143; 16,229
monoglyme, ethylene glycol dimethyl ether, glyme, dimethyl cellosolve, egdme, ethane, 1,2-dimethoxy, dimethoxyethane, 2,5-dioxahexane, glycol dimethyl ether, ethylene dimethyl ether
≥99%
128-37-0
Pass Test
99%
Vapor Pressure: 64hPa at 20°C
50ppm max.
15, 3245
90.12
Extra Dry
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if inhaled.
May damage fertility. May damage the unborn child.
Highly flammable liquid and vapor.
May form explosive peroxides.

GHS P Statement:
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Keep container tightly closed.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

missing translation for 'einecsNumber' : 203-794-9

missing translation for 'rtecsNumber' : KI1451000

missing translation for 'tsca' : TSCA

Product Identifier
  • Ethylene glycol dimethyl ether

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Flammable liquid Category 2
  • Reproductive toxicity Category 1B
  • Skin corrosion/irritation Category 2

Hazard Statement
  • H360FD-May damage fertility. May damage the unborn child.
  • H315-Causes skin irritation.
  • H332-Harmful if inhaled.
  • H225-Highly flammable liquid and vapour.
  • EUH019-May form explosive peroxides.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.