missing translation for 'onlineSavingsMsg'
Learn More

Ethylene Glycol, Fisher BioReagents™

37.00 € - 65.15 €

Chemical Identifiers

CAS 107-21-1
Molecular Formula C2H6O2
Molecular Weight (g/mol) 62.068
InChI Key LYCAIKOWRPUZTN-UHFFFAOYSA-N
Synonym ethylene glycol, 1,2-ethanediol, glycol, monoethylene glycol, 1,2-dihydroxyethane, 2-hydroxyethanol, glycol alcohol, ethylene alcohol, fridex, tescol
PubChem CID 174
ChEBI CHEBI:30742
IUPAC Name ethane-1,2-diol
SMILES C(CO)O
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10011073
View Documents Promotion Details
Fisher BioReagents
BP230-1
1 L Glass Bottle
37.00 €
1 L
Please to purchase this item. Need a web account? Register with us today!
10532595
View Documents Promotion Details
Fisher BioReagents
BP230-4
4 L Glass Bottle
65.15 €
4 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Ethylene Glycol

Due to its property as an antifreeze, Ethylene Glycol may be used for the storage of enzymes at low temperatures.
Specifications

Chemical Identifiers

107-21-1
62.068
ethylene glycol, 1,2-ethanediol, glycol, monoethylene glycol, 1,2-dihydroxyethane, 2-hydroxyethanol, glycol alcohol, ethylene alcohol, fridex, tescol
CHEBI:30742
C(CO)O
C2H6O2
LYCAIKOWRPUZTN-UHFFFAOYSA-N
174
ethane-1,2-diol

Specifications

5.5 to 7.5
107-21-1
>500°C
196°C
Viscous Liquid
C2H6O2
HOCH2CH2OH
21 cP at 20°C
15, 3852
LYCAIKOWRPUZTN-UHFFFAOYSA-N
ethane-1,2-diol
174
62.06
Ethylene Glycol
-13°C
Colorless
1.113g/cm³
Pass Test
≥99 %
0.002mEq/g max.
0.12mmHg at 20°C
0.5% max.
ethylene glycol, 1,2-ethanediol, glycol, monoethylene glycol, 1,2-dihydroxyethane, 2-hydroxyethanol, glycol alcohol, ethylene alcohol, fridex, tescol
C(CO)O
62.068
CHEBI:30742
≥99%
Videos
Safety and Handling

Safety and Handling

WARNING!

Emergency Overview
Harmful if swallowed. May cause skin, eye, and respiratory tract irritation. Hygroscopic. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Get medical attention immediately if symptoms occur.

NFPA

Health:2
Flammability:1
Instability:0

Recommended Storage : RT

Product Identifier
  • Ethylene glycol

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Specific target organ toxicity after repeated exposure Category 2

Hazard Statement
  • H373-May cause damage to organs.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /

SDS
Documents

Documents

Product Certifications