Learn More
Ethylene glycol-O,O'-bis(2-aminoethyl)-N,N,N',N'-tetraacetic acid, 97%, Thermo Scientific Chemicals
49.30 € - 555.00 €
Chemical Identifiers
CAS | 67-42-5 |
---|---|
Molecular Formula | C14H24N2O10 |
Molecular Weight (g/mol) | 380.35 |
MDL Number | MFCD00004291 |
InChI Key | DEFVIWRASFVYLL-UHFFFAOYSA-N |
Synonym | egta, egtazic acid, gedta, ethylenebis oxyethylenenitrilo tetraacetic acid, ebonta, 6,9-dioxa-3,12-diazatetradecanedioic acid, 3,12-bis carboxymethyl, 1,2-bis 2-bis carboxymethyl amino ethoxy ethane, ethylene glycol tetraacetic acid, h4egta, egtazic acid usan:inn |
PubChem CID | 6207 |
ChEBI | CHEBI:30740 |
IUPAC Name | 2-[2-[2-[2-[bis(carboxymethyl)amino]ethoxy]ethoxy]ethyl-(carboxymethyl)amino]acetic acid |
SMILES | C(COCCOCCN(CC(=O)O)CC(=O)O)N(CC(=O)O)CC(=O)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11443097
|
Thermo Scientific Alfa Aesar
A16086.09 |
10 g |
49.30 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11453097
|
Thermo Scientific Alfa Aesar
A16086.18 |
50 g |
138.00 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11463097
|
Thermo Scientific Alfa Aesar
A16086.30 |
250 g |
555.00 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Ethylene glycol-O,O'-bis(2-aminoethyl)-N,N,N',N'-tetraacetic acid is a chelating agent used for making complexes, especially for calcium ions, which help to determine calcium in the presence of magnesium. It can be used experimentally for treating animals possessing cerium poisoning. It is useful in the separation of thorium from monazite. It also acts as a buffer and is used in protein purification.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsEthylene glycol-O,O′-bis(2-aminoethyl)-N,N,N′,N′-tetraacetic acid is a chelating agent used for making complexes, especially for calcium ions, which help to determine calcium in the presence of magnesium. It can be used experimentally for treating animals possessing cerium poisoning. It is useful in the separation of thorium from monazite. It also acts as a buffer and is used in protein purification.
Solubility
Soluble in 0.1M sodium hydroxide solution. Slightly soluble in water.
Notes
Incompatible with strong oxidizing agents, alkali, alkali earth metal and many reactive organic and inorganic chemicals.
Chemical Identifiers
67-42-5 | |
380.35 | |
DEFVIWRASFVYLL-UHFFFAOYSA-N | |
6207 | |
2-[2-[2-[2-[bis(carboxymethyl)amino]ethoxy]ethoxy]ethyl-(carboxymethyl)amino]acetic acid |
C14H24N2O10 | |
MFCD00004291 | |
egta, egtazic acid, gedta, ethylenebis oxyethylenenitrilo tetraacetic acid, ebonta, 6,9-dioxa-3,12-diazatetradecanedioic acid, 3,12-bis carboxymethyl, 1,2-bis 2-bis carboxymethyl amino ethoxy ethane, ethylene glycol tetraacetic acid, h4egta, egtazic acid usan:inn | |
CHEBI:30740 | |
C(COCCOCCN(CC(=O)O)CC(=O)O)N(CC(=O)O)CC(=O)O |
Specifications
67-42-5 | |
C14H24N2O10 | |
1717370 | |
egta, egtazic acid, gedta, ethylenebis oxyethylenenitrilo tetraacetic acid, ebonta, 6,9-dioxa-3,12-diazatetradecanedioic acid, 3,12-bis carboxymethyl, 1,2-bis 2-bis carboxymethyl amino ethoxy ethane, ethylene glycol tetraacetic acid, h4egta, egtazic acid usan:inn | |
DEFVIWRASFVYLL-UHFFFAOYSA-N | |
2-[2-[2-[2-[bis(carboxymethyl)amino]ethoxy]ethoxy]ethyl-(carboxymethyl)amino]acetic acid | |
6207 | |
380.35 | |
Ethylene glycol-O,O'-bis(2-aminoethyl)-N,N,N',N'-tetraacetic acid |
∼242°C (decomposition) | |
MFCD00004291 | |
14,3529 | |
Soluble in 0.1M sodium hydroxide solution. Slightly soluble in water. | |
C(COCCOCCN(CC(=O)O)CC(=O)O)N(CC(=O)O)CC(=O)O | |
380.35 | |
CHEBI:30740 | |
97% |
Safety and Handling
missing translation for 'einecsNumber' : 200-651-2
missing translation for 'rtecsNumber' : AH3760000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only