missing translation for 'onlineSavingsMsg'
Learn More

Ethylene glycol, Reagent Grade, ≥99%, Honeywell™

Product Code. 15680960
Click to view available options
Quantity:
500 mL
1 L
2.5 L
5 L
6 x 500 mL
20 L
30 kg
4 x 2.5 L
225 kg
Packaging:
Metal Drum
Poly Bottle
Stainless Steel Can
This item is not returnable. View return policy
This item is not returnable. View return policy

Reagent Grade,≥99%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107-21-1
Molecular Formula C2H6O2
Molecular Weight (g/mol) 62.068
MDL Number MFCD00002885
InChI Key LYCAIKOWRPUZTN-UHFFFAOYSA-N
Synonym ethylene glycol, 1,2-ethanediol, glycol, monoethylene glycol, 1,2-dihydroxyethane, 2-hydroxyethanol, glycol alcohol, ethylene alcohol, fridex, tescol
PubChem CID 174
ChEBI CHEBI:30742
IUPAC Name ethane-1,2-diol
SMILES C(CO)O

Specifications

Melting Point -13°C (literature)
CAS Max % 100.0000%
Density 1.113 g/mL at 25 °C(lit.)
Boiling Point 197°C
Quantity 30 kg
Flash Point 111°C
Packaging Stainless Steel Can
Linear Formula HOCH2CH2OH
Beilstein 505945
Formula Weight 62.07
Percent Purity >99%
Grade Reagent
Chemical Name or Material Ethylene glycol, Reagent Grade, ≥99%
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'