missing translation for 'onlineSavingsMsg'
Learn More
Ethylene glycol, technical Product Code.: 13486369

Ethylene glycol, technical

Product Code. 13486369
5 L, Plastic Drum
Click to view available options
Quantity:
5 L
Packaging:
Plastic Drum
This item is not returnable. View return policy

Product Code. 13486369

Brand: Thermo Scientific Acros 444230050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 107-21-1 | C2H6O2 | 62.068 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Melting Point -13.0°C
CAS 107-21-1
Boiling Point 196.0°C to 198.0°C
Quantity 5 L
Flash Point 116°C
Packaging Plastic Drum
Refractive Index 1.4300 to 1.4330
Specific Gravity 1.113
Molecular Formula C2H6O2
Linear Formula HOCH2CH2OH
Viscosity 21 mPa.s (20°C)
Synonym ethylene glycol, 1,2-ethanediol, glycol, monoethylene glycol, 1,2-dihydroxyethane, 2-hydroxyethanol, glycol alcohol, ethylene alcohol, fridex, tescol
Solubility Information Solubility in water: miscible. Other solubilities: miscible with acetic acid, acetone, glycerol,, pyridine, lower aliphatic alcohols,, aldehydes and ketones, slightly soluble in ether, practically insoluble in benzene and hydrocarbons
InChI Key LYCAIKOWRPUZTN-UHFFFAOYSA-N
SMILES C(CO)O
IUPAC Name ethane-1,2-diol
Molecular Weight (g/mol) 62.068
PubChem CID 174
ChEBI CHEBI:30742
Formula Weight 62.06
Grade Technical
Chemical Name or Material Ethylene Glycol
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0464404 Certificate TypeCertificate of Analysis Date10/07/2024 Catalog Number444230010,444230050
Lot NumberA0462682 Certificate TypeCertificate of Analysis Date09/05/2024 Catalog Number444230010,444230050
Lot NumberA0461770 Certificate TypeCertificate of Analysis Date09/04/2024 Catalog Number444230010,444230050
Lot NumberA0461218 Certificate TypeCertificate of Analysis Date20/03/2024 Catalog Number444230010,444230050
Lot NumberA0460830 Certificate TypeCertificate of Analysis Date08/03/2024 Catalog Number444230010,444230050
compliance-icons compliance-icons
Product Identifier
  • Ethylene glycol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H373-May cause damage to organs.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
Supplemental information
  • MIXTURE LIST-Contains : Ethylene glycol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ethylene glycol, technical >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.