missing translation for 'onlineSavingsMsg'
Learn More
Ethylenediamine, 99+%, Thermo Scientific Chemicals Product Code.: 10593191

Ethylenediamine, 99+%, Thermo Scientific Chemicals

Product Code. 10593191 Shop All Thermo Scientific Products
250 mL, Glass bottle
Click to view available options
Quantity:
250 mL
1 L
2.5 L
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10593191

Brand: Thermo Scientific 220422500

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107-15-3
Molecular Formula C2H8N2
Molecular Weight (g/mol) 60.10
MDL Number MFCD00008204
InChI Key PIICEJLVQHRZGT-UHFFFAOYSA-N
Synonym ethylenediamine, 1,2-ethanediamine, 1,2-diaminoethane, ethylene diamine, ethylenediamine, dimethylenediamine, edamine, 1,2-ethylenediamine, aethaldiamin, aethylenediamin
PubChem CID 3301
ChEBI CHEBI:30347
IUPAC Name ethane-1,2-diamine
SMILES NCCN

Specifications

Melting Point 11.0°C
Density 0.8980g/mL
Boiling Point 117.0°C to 118.0°C
Flash Point 38°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Linear Formula H2NCH2CH2NH2
Refractive Index 1.4555 to 1.4575
Quantity 250 mL
Beilstein 04, 230
Fieser 01,372; 04,231; 13,157
Merck Index 15, 3849
Specific Gravity 0.898
Solubility Information Solubility in water: freely soluble. Other solubilities: freely soluble in alcohol, slightly soluble in ether, soluble in benzene
Viscosity 1.6 mPa.s (20°C)
Formula Weight 60.1
Percent Purity 99+%
Grade Extra Pure
Physical Form Liquid, Fuming In Air
Chemical Name or Material Ethylenediamine
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0464136 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number220422500,220420025,220420010
Lot NumberA0464136 Certificate TypeCertificate of Analysis Date02/07/2024 Catalog Number220422500,220420025,220420010
Lot NumberA0454475 Certificate TypeCertificate of Analysis Date29/04/2024 Catalog Number220422500,220420025,220420010
Lot NumberA0446930 Certificate TypeCertificate of Analysis Date02/05/2023 Catalog Number220422500,220420025,220420010
Lot NumberA0404297 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number220422500,220420025,220420010
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Ethylenediamine
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
  • Acute toxicity Category 3
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H311-Toxic in contact with skin.
  • H332-Harmful if inhaled.
  • H314-Causes severe skin burns and eye damage.
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ethylenediamine, 99+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.