missing translation for 'onlineSavingsMsg'
Learn More

Ethylenediamine Tetraacetate, DEPC-Treated Solution (0.5M, pH 8.0), Fisher BioReagents

Product Code. p-25771
Click to view available options
Quantity:
100 mL
500 mL
1 L
Packaging:
Nalgene Poly Bottle
Poly Bottle
Unit Size:
1 L
100mL
500mL
This item is not returnable. View return policy

Product Code. 10572595

Brand: Fisher BioReagents BP2483100

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
EDTA, DEPC-Treated solution is ideal for RNA work.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60-00-4, 1310-73-2, 1609-47-8
Molecular Formula C10H16N2O8
Molecular Weight (g/mol) 292.24
MDL Number MFCD00003541
InChI Key KCXVZYZYPLLWCC-UHFFFAOYSA-N
Synonym edta, edetic acid, ethylenediaminetetraacetic acid, edathamil, versene, endrate, havidote, titriplex, edta acid, sequestrol
PubChem CID 6049
ChEBI CHEBI:42191
IUPAC Name 2-[2-[bis(carboxymethyl)amino]ethyl-(carboxymethyl)amino]acetic acid
SMILES OC(=O)CN(CCN(CC(O)=O)CC(O)=O)CC(O)=O

Specifications

Color Colorless
Concentration 0.475 to 0.525M
Merck Index 15, 3565
Identification Pass Test
Packaging Poly Bottle
pH 8.0
Quantity 100 mL
Physical Form Liquid
Chemical Name or Material Ethylenediamine Tetraacetate, DEPC-Treated Solution, 0.5M, pH 8.0
compliance-icons compliance-icons
Product Identifier
  • Ethylenediamine Tetraacetate, DEPC-Treated Solution (0.5M, pH 8.0)
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H373-May cause damage to organs.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Ethylenediamine tetraacetic acid (EDTA),Diethyl pyrocarbonate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.