missing translation for 'onlineSavingsMsg'
Learn More
Ethylenediaminetetraacetic Acid (0.5M Solution/pH 8.0), Fisher BioReagents Product Code.: 10628203

Ethylenediaminetetraacetic Acid (0.5M Solution/pH 8.0), Fisher BioReagents

Product Code. 10628203
1 L, Nalgene Poly Bottle
Click to view available options
Quantity:
100 mL
500 mL
1 L
20 L
Packaging:
Nalgene Poly Bottle
Poly Bottle
Poly CUBE
This item is not returnable. View return policy

Product Code. 10628203

Brand: Fisher BioReagents BP24821

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy
EDTA is used extensively in molecular biology to minimize metal ion impurities in reaction buffers. Ideal for DNA work.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60-00-4, 7732-18-5
Molecular Formula C10H16N2O8
Molecular Weight (g/mol) 292.24
MDL Number MFCD00003541
InChI Key KCXVZYZYPLLWCC-UHFFFAOYSA-N
Synonym edta, edetic acid, ethylenediaminetetraacetic acid, edathamil, versene, endrate, havidote, titriplex, edta acid, sequestrol
PubChem CID 6049
ChEBI CHEBI:42191
IUPAC Name 2-[2-[bis(carboxymethyl)amino]ethyl-(carboxymethyl)amino]acetic acid
SMILES OC(=O)CN(CCN(CC(O)=O)CC(O)=O)CC(O)=O

Specifications

Color Colorless
Linear Formula (HOOCCH2)2NCH2CH2N(CH2COOH)2
Concentration 0.475 to 0.525M
Merck Index 15, 3565
Formula Weight 292.25
Identification Pass Test
Packaging Nalgene Poly Bottle
pH 8.0
Quantity 1 L
Physical Form Liquid
Chemical Name or Material Ethylenediamine Tetraacetic Acid, 0.5 M Solution, pH 8.0 (for DNA Work)
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Ethylenediaminetetraacetic Acid (0.5M Solution/pH 8.0)
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H373-May cause damage to organs.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ethylenediaminetetraacetic Acid (0.5M Solution/pH 8.0), Fisher BioReagents >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.