missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals Ethylenediaminetetraacetic acid, disodium salt dihydrate, 99+%, for analysis Product Code.: 10416790

Thermo Scientific Chemicals Ethylenediaminetetraacetic acid, disodium salt dihydrate, 99+%, for analysis

Product Code. 10416790
500 g, Plastic bottle
Click to view available options
Quantity:
100 g
500 g
1 kg
5 kg
Packaging:
Plastic bottle
Plastic drum
This item is not returnable. View return policy

Product Code. 10416790

Brand: Thermo Scientific Chemicals 147855000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Biological buffer, Chelation/Complexation reagent

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Biological buffer, Chelation/Complexation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6381-92-6
Molecular Formula C10H18N2Na2O10
Molecular Weight (g/mol) 372.24
MDL Number MFCD00150037,MFCD00003541
InChI Key OVBJJZOQPCKUOR-UHFFFAOYSA-L
Synonym edta disodium salt, cal-ex decalcifier, buffer solution, ph 10.00, sodium di ethylenediamine tetraacetate dihydrate, ethylenediamine tetraacetic acid, disodium salt dihydrate, ethylenediamine tetraacetic acid, disodium salt, standard solution, sodium di ethylenediamine tetraacetate standard solution, ethylenedinitrilo tetraacetic acid disodium, dihydrate, reagent, acs
PubChem CID 44120005
IUPAC Name disodium;2-[2-[bis(carboxymethyl)amino]ethyl-(carboxylatomethyl)amino]acetate;dihydrate
SMILES O.O.[Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O

Specifications

Melting Point 252.0°C
Color White
pH 4.0 to 6.0 (1% soln. at 20°C)
Loss on Drying 11.4% max. (200°C, 3 hrs)
Infrared Spectrum Authentic
Assay Percent Range 99.0 to 101% (On dry substance)
Packaging Plastic bottle
Quantity 500 g
Merck Index 15, 3565
Solubility Information Solubility in water: freely soluble. Other solubilities: slightly soluble in alcohol (95%),insoluble in chloroform and ether
Formula Weight 372.23
Percent Purity 99+%
Grade Analysis
Physical Form Fine Crystalline Powder
Chemical Name or Material Ethylenediaminetetraacetic acid, disodium salt dihydrate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0465869 Certificate TypeCertificate of Analysis Date06/09/2024 Catalog Number147850050,147855000,147850010,147851000
Lot NumberA0460697 Certificate TypeCertificate of Analysis Date08/03/2024 Catalog Number147850050,147855000,147850010,147851000
Lot NumberA0455923 Certificate TypeCertificate of Analysis Date28/08/2023 Catalog Number147850050,147855000,147850010,147851000
Lot NumberA0453477 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number147850050,147855000,147850010,147851000
Lot NumberA0401588 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number147850050,147855000,147850010,147851000
compliance-icons compliance-icons
Product Identifier
  • Ethylenediaminetetraacetic acid, disodium salt dihydrate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H373-May cause damage to organs.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals Ethylenediaminetetraacetic acid, disodium salt dihydrate, 99+%, for analysis >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.