missing translation for 'onlineSavingsMsg'
Learn More
Ethylenediaminetetraacetic Acid, Disodium Salt Dihydrate, Extra Pure, SLR, Fisher Chemical™ Product Code.: 10716481
compliance-icons compliance-icons

Ethylenediaminetetraacetic Acid, Disodium Salt Dihydrate, Extra Pure, SLR, Fisher Chemical™

Product Code. 10716481
1 kg, Plastic powder jar
Click to view available options
Quantity:
100 g
250 g
500 g
1 kg
5 kg
Packaging:
Plastic keg
Plastic powder jar

Product Code. 10716481

Brand: Fisher Chemical D/0650/60

Please to purchase this item. Need a web account? Register with us today!

Chemical Identifiers

CAS 6381-92-6
Molecular Formula C10H18N2Na2O10
Molecular Weight (g/mol) 372.24
MDL Number 150037
InChI Key OVBJJZOQPCKUOR-UHFFFAOYSA-L
IUPAC Name disodium 2-({2-[(carboxylatomethyl)(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetate dihydrate
SMILES O.O.[Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O

Specifications

Melting Point 252°C
Color White
pH 4 to 6
Physical Form Solid
Quantity 1 kg
Formula Weight 372.23g/mol
Packaging Plastic powder jar
compliance-icons compliance-icons
Product Identifier
  • Ethylenediaminetetraacetic acid, disodium salt dihydrate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H373-May cause damage to organs.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ethylenediaminetetraacetic Acid, Disodium Salt Dihydrate, Extra Pure, SLR, Fisher Chemical™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.