missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethylenediaminetetraacetic Acid Disodium Salt Solution 0.01M (0.02N), NIST Standard Solution ready to use, for volumetric analysis, Fisher Chemical™
Click to view available options
Quantity:
1 L
2.5 L
10 L
Packaging:
Bag in box
HDPE plastic bottle
Description
Ethylenediaminetetraacetic Acid Disodium Salt Solution 0.01M (0.02N), NIST Standard Solution ready to use, for volumetric analysis

Specifications
Specifications
Color | Colorless |
Physical Form | Liquid |
Quantity | 2.5 L |
Packaging | HDPE plastic bottle |
Safety and Handling
Product Identifier
- EDTA (Diaminoethanetetra-acetic acid, disodium salt solution 0.1 M) (0.2N)
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction