missing translation for 'onlineSavingsMsg'
Learn More

Ethylenediaminetetraacetic Acid Disodium Salt Solution, 0.2M, Honeywell Fluka™

Product Code. 15654800
Click to view available options
Quantity:
1 L
5 L
10 L
6 x 1 L
Packaging:
Cubitainer
Poly Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Volumetric, 0.2 M EDTA-Na2, for complexometry

Honeywell´s high-quality titration products cover the complete titration workflow and include:

  • Solutions for volumetric titration (acids, bases, or salts)
  • Buffers available as concentrates or ready-to-use

 

  • Reagents for complexometry:
    • Aminopolycarboxylic acids (EDTA/NTA analogs)
    • Masking agents
  • Indicators for volumetric and complexometric titrations

 


 

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 139-33-3
Molecular Formula C10H14N2Na2O8
MDL Number MFCD00070672,MFCD00003541,MFCD00070672,MFCD00150037
UN Number NONH for all modes of transport
Beilstein 3822669
Synonym ethylenediaminetetraacetic acid disodium salt, edta na2, disodium 2-2-bis carboxymethyl amino ethyl-carboxymethyl amino acetic acid
InChI Key ZGTMUACCHSMWAC-UHFFFAOYSA-L
SMILES [Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O
Molecular Weight (g/mol) 336.21
PubChem CID 57339238
ChEBI CHEBI:64734
Chemical Name or Material Ethylenediaminetetraacetic acid disodium salt solution
Show More Show Less

Specifications

Linear Formula [CH2N(CH2CO2Na)CH2CO2H]2
IUPAC Name disodium 2-({2-[(carboxylatomethyl)(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetate
Formula Weight 336.21
Packaging Poly Bottle
Quantity 10 L
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'