Learn More
EDTA disodium salt solution with zinc complex added (solution B), 1 ml solution = 1 German degree of hardness in 100 ml of water, for complexometry, Honeywell Fluka™
1 ml solution = 1 German degree of hardness in 100 ml of water, for complexometry
Brand:  Honeywell-Fluka 34544-1L
 View more versions of this product
Product Code. 15614260
  • 77.46 € / 1 L
Explore more special offers
Description
Description
Honeywell´s high-quality titration products cover the complete titration workflow and include:
  • Solutions for volumetric titration (acids, bases, or salts)
  • Buffers available as concentrates or ready-to-use
  • Reagents for complexometry:
    • Aminopolycarboxylic acids (EDTA/NTA analogs)
    • Masking agents
  • Indicators for volumetric and complexometric titrations
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Ethylenediaminetetraacetic acid disodium salt solution with zinc complex added (solution B)
0.7, 0.02
NONH for all modes of transport
[Cl-].[Cl-].[Zn++]
136.28
1 L
7646-85-7, 60-00-4, 7732-18-5
Cl2Zn
JIAARYAFYJHUJI-UHFFFAOYSA-L
zinc(2+) dichloride
Poly Bottle