missing translation for 'onlineSavingsMsg'
Learn More

EDTA disodium salt solution with zinc complex added (solution B), 1 ml solution = 1 German degree of hardness in 100 ml of water, for complexometry, Honeywell Fluka™

Product Code. 15644260
Click to view available options
Quantity:
1 L
5 L
6 x 500 mL
10 L
6 x 1 L
Packaging:
Cubitainer
Poly Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

1 ml solution = 1 German degree of hardness in 100 ml of water, for complexometry

Honeywell´s high-quality titration products cover the complete titration workflow and include:

  • Solutions for volumetric titration (acids, bases, or salts)
  • Buffers available as concentrates or ready-to-use

 

  • Reagents for complexometry:
    • Aminopolycarboxylic acids (EDTA/NTA analogs)
    • Masking agents
  • Indicators for volumetric and complexometric titrations

 


 

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7646-85-7, 60-00-4, 7732-18-5
UN Number NONH for all modes of transport

Specifications

CAS Max % 0.02, 0.70
Molecular Formula Cl2Zn
InChI Key JIAARYAFYJHUJI-UHFFFAOYSA-L
SMILES [Cl-].[Cl-].[Zn++]
IUPAC Name zinc(2+) dichloride
Molecular Weight (g/mol) 136.28
Packaging Cubitainer
Quantity 5 L
Chemical Name or Material Ethylenediaminetetraacetic acid disodium salt solution with zinc complex added (solution B)
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'