missing translation for 'onlineSavingsMsg'
Learn More

EDTA disodium salt solution with zinc complex added (solution B), 1 ml solution = 1 German degree of hardness in 100 ml of water, for complexometry, Honeywell Fluka™

Product Code. 15654260
Click to view available options
Quantity:
1 L
5 L
6 x 500 mL
10 L
6 x 1 L
Packaging:
Cubitainer
Poly Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

1 ml solution = 1 German degree of hardness in 100 ml of water, for complexometry

Honeywell´s high-quality titration products cover the complete titration workflow and include:

  • Solutions for volumetric titration (acids, bases, or salts)
  • Buffers available as concentrates or ready-to-use

 

  • Reagents for complexometry:
    • Aminopolycarboxylic acids (EDTA/NTA analogs)
    • Masking agents
  • Indicators for volumetric and complexometric titrations

 


 

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7646-85-7, 60-00-4, 7732-18-5
UN Number NONH for all modes of transport

Specifications

Boiling Point 100°C
CAS Min % 0.7000%
CAS Max % >99.0000%
Molecular Formula C10H16N2O8
Linear Formula C6H12
MDL Number MFCD00003541
Density 1.000 g/cm3
Synonym edta, edetic acid, ethylenediaminetetraacetic acid, edathamil, versene, endrate, havidote, titriplex, edta acid, sequestrol
InChI Key KCXVZYZYPLLWCC-UHFFFAOYSA-N
SMILES OC(=O)CN(CCN(CC(O)=O)CC(O)=O)CC(O)=O
IUPAC Name 2-({2-[bis(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetic acid
Molecular Weight (g/mol) 292.24
PubChem CID 6049
ChEBI CHEBI:42191
Packaging Poly Bottle
Quantity 6 x 1 L
Physical Form Buffer/Volumetric Solutions
Chemical Name or Material EDTA disodium salt solution with zinc complex added (solution B), 1 ml solution = 1 German degree of hardness in 100 ml of water, for complexometry
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'