missing translation for 'onlineSavingsMsg'
Learn More

Ethylenediaminetetraacetic Acid, Honeywell Fluka™

Product Code. 15690680
Click to view available options
Quantity:
500 g
1 kg

for complexometry,≥99.0%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60-00-4
Molecular Formula C10H16N2O8
Molecular Weight (g/mol) 292.24
MDL Number MFCD00003541
InChI Key KCXVZYZYPLLWCC-UHFFFAOYSA-N
Synonym edta, edetic acid, ethylenediaminetetraacetic acid, edathamil, versene, endrate, havidote, titriplex, edta acid, sequestrol
PubChem CID 6049
ChEBI CHEBI:42191
SMILES OC(=O)CN(CCN(CC(O)=O)CC(O)=O)CC(O)=O

Specifications

Ignition Residue ≤0.1% (as SO4)
Packaging Plastic Bottle
Linear Formula (HO2CCH2)2NCH2CH2N(CH2CO2H)2
Quantity 1 kg
Beilstein 1716295
Solubility Information Soluble in water (0.4g/L at 20°C)
IUPAC Name 2-[2-[bis(carboxymethyl)amino]ethyl-(carboxymethyl)amino]acetic acid
Formula Weight 292.24g/mol
Percent Purity ≥99%
Chemical Name or Material Ethylenediaminetetraacetic Acid
compliance-icons
Product Identifier
  • Ethylenediaminetetraacetic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Ethylenediaminetetraacetic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'