missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Ethylenediaminetetraacetic acid, trisodium salt hydrate, 98%

Product Code. 10081490 Shop All Thermo Scientific Chemicals Products
Click to view available options
:
500g; Plastic bottle
This item is not returnable. View return policy

Product Code. 10081490

Brand: Thermo Scientific Chemicals undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 85715-60-2 | C10H13N2Na3O8 | 358.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chelation/Complexation reagent
TRUSTED_SUSTAINABILITY

Specifications

CAS 85715-60-2
Melting Point 300.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (corrected for water) (Complexometry)
Packaging Plastic bottle
Molecular Formula C10H13N2Na3O8
Linear Formula (NaO2CCH2)2NCH2CH2N(CH2CO2Na)CH2CO2H·xH2O
MDL Number MFCD00149675
Quantity 500 g
Synonym trisodium hydrogen ethylenediaminetetraacetate hydrate, tricon bw, unii-zmn5a047nl, endrate, edta triasodium salt, zmn5a047nl, ethylenediaminetetraacetic acid trisodium salt hydrate, sequestrene sodium 3, trisodium edta monohydrate, edetate trisodium monohydrate
Solubility Information Solubility in water: soluble.
InChI Key QZKRHPLGUJDVAR-UHFFFAOYSA-K
SMILES [Na+].[Na+].[Na+].OC(=O)CN(CCN(CC([O-])=O)CC([O-])=O)CC([O-])=O
IUPAC Name trisodium;2-[2-[bis(carboxylatomethyl)amino]ethyl-(carboxymethyl)amino]acetate;hydrate
Molecular Weight (g/mol) 358.19
PubChem CID 16211179
Formula Weight 394.22
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Ethylenediaminetetraacetic acid, trisodium salt hydrate, 98%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Ethylenediaminetetraacetic acid, trisodium salt hydrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H318-Causes serious eye damage.
  • H332-Harmful if inhaled.
  • H373-May cause damage to organs.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Ethylenediaminetetraacetic acid, trisodium salt hydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.