missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethylenediaminetetraacetic Acid Trisodium Salt Solution 0.1M (0.2N), NIST Standard Concentrate, for volumetric analysis, Fisher Chemical™
Click to view available options
Quantity:
1 AMP
6 AMP
Specifications
Specifications
Color | Colorless |
pH | 4 to 5 |
Physical Form | Liquid |
Quantity | 1 AMP |
Packaging | Ampoule |
Safety and Handling
Product Identifier
- EDTA (Diaminoethanetetra-acetic acid, disodium salt solution 0.1 M) (0.2N)
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'