missing translation for 'onlineSavingsMsg'
Learn More
Ethylhydrocupreine hydrochloride, 97%, Thermo Scientific Chemicals Product Code.: 11470167

Ethylhydrocupreine hydrochloride, 97%, Thermo Scientific Chemicals

Product Code. 11470167
100 mg
Click to view available options
Quantity:
100 mg
500 mg
This item is not returnable. View return policy

Product Code. 11470167

Brand: Thermo Scientific Alfa Aesar B21627.MC

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3413-58-9
Molecular Formula C21H29ClN2O2
Molecular Weight (g/mol) 376.925
MDL Number MFCD00135594
InChI Key QNRATNLHPGXHMA-XZHTYLCXSA-N
Synonym ethylhydrocupreine hydrochloride, optoquin hydrochloride, ethylhydrocupreine hcl, optochin hydrochloride, unii-s3n0jb4fc7, neumolisina, numoquin hydrochloride, s3n0jb4fc7, optoquinhydrochloride, optochin
PubChem CID 16219340
IUPAC Name (R)-(6-ethoxyquinolin-4-yl)-[(2S,4S,5R)-5-ethyl-1-azabicyclo[2.2.2]octan-2-yl]methanol;hydrochloride
SMILES CCC1CN2CCC1CC2C(C3=C4C=C(C=CC4=NC=C3)OCC)O.Cl

Specifications

Melting Point ∼240°C (decomposition)
Quantity 100 mg
Sensitivity Light sensitive
Formula Weight 374.91
Percent Purity 97%
Chemical Name or Material Ethylhydrocupreine hydrochloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ethylhydrocupreine hydrochloride, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.