missing translation for 'onlineSavingsMsg'
Learn More

Ethylmagnesium bromide, 3M in diethyl ether, AcroSeal™, Thermo Scientific Chemicals

Product Code. 10601425
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass bottle
This item is not returnable. View return policy
This item is packed in AcroSeal™, the industry-leading packaging for air and moisture sensitive reagents.
Find out more about AcroSeal™ packaging.
 
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
This item is not returnable. View return policy
This item is packed in AcroSeal™, the industry-leading packaging for air and moisture sensitive reagents.
Find out more about AcroSeal™ packaging.
 
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 925-90-6, 60-29-7
Molecular Formula C2H5BrMg
Molecular Weight (g/mol) 133.27
MDL Number MFCD00000043
InChI Key TWTWFMUQSOFTRN-UHFFFAOYSA-M
Synonym ethylmagnesium bromide, ethyl magnesium bromide, magnesium, bromoethyl, ethylmagnesiumbromide, bromoethylmagnesium, ethylmagnesium bromide solution, 1.0 m in thf, ethylmagnesium bromide solution, 3.0 m in diethyl ether, etmgbr, ethylmagnesiumbromid, ethylmagnesium bromid, grignard reagent
PubChem CID 101914
IUPAC Name magnesium;ethane;bromide
SMILES CC[Mg]Br

Specifications

Color Brown
Density 1.0300g/mL
Flash Point −40°C
Packaging AcroSeal™ Glass bottle
Linear Formula C2H5MgBr
Quantity 100 mL
Fieser 02,205
Specific Gravity 1.03
Formula Weight 133.27
Physical Form Liquid
Chemical Name or Material Ethylmagnesium bromide
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Ethylmagnesium bromide, 3M in Diethyl ether
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
  • Substance which\, in contact with water\, emits flammable gases Category 1
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H336-May cause drowsiness or dizziness.
  • H302-Harmful if swallowed.
  • H260-In contact with water releases flammable gases which may ignite spontaneously.
  • H224-Extremely flammable liquid and vapour.
  • EUH014-Reacts violently with water.
  • EUH019-May form explosive peroxides.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P223-Do not allow contact with water.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'