missing translation for 'onlineSavingsMsg'
Learn More
Ethylmagnesium chloride, 2.7M (25 wt.%) solution in THF, AcroSeal™, Thermo Scientific Chemicals Product Code.: 10515223

Ethylmagnesium chloride, 2.7M (25 wt.%) solution in THF, AcroSeal™, Thermo Scientific Chemicals

Product Code. 10515223
100 mL, AcroSeal™ Glass bottle
Click to view available options
Quantity:
100 mL
800 mL
2.4 L
Packaging:
AcroSeal™ Glass bottle
This item is not returnable. View return policy
This item is packed in AcroSeal™, the industry-leading packaging for air and moisture sensitive reagents.
Find out more about AcroSeal™ packaging.
 
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

Product Code. 10515223

Brand: Thermo Scientific Acros 252571000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
This item is packed in AcroSeal™, the industry-leading packaging for air and moisture sensitive reagents.
Find out more about AcroSeal™ packaging.
 
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2386-64-3, 109-99-9
Molecular Formula C2H5ClMg
Molecular Weight (g/mol) 88.82
MDL Number MFCD00000472
InChI Key ZWNJJACKPNTXHY-UHFFFAOYSA-M
Synonym ethylmagnesium chloride, chloroethylmagnesium, ethylmagnesiumchloride, magnesium, chloroethyl, etmgcl, chloro ethyl magnesium, ethyl magnesium chloride, chloroethyl-magnesiu, ethyl magnesiumchloride, ethylmagnesium chloride solutionShow MoreShow Less
PubChem CID 75432
IUPAC Name magnesium;ethane;chloride
SMILES CC[Mg]Cl

Specifications

Color Brown
Density 1.0000g/mL
Packaging AcroSeal™ Glass bottle
Linear Formula C2H5MgCl
Quantity 100 mL
Specific Gravity 1
Solubility Information Solubility in water: reacts
Formula Weight 88.83
Percent Purity 24 to 26%
Physical Form Liquid
Chemical Name or Material Ethylmagnesium chloride
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0467260 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number252571000,252578000
Lot NumberA0461132 Certificate TypeCertificate of Analysis Date18/03/2024 Catalog Number252571000,252578000
Lot NumberA0393543 Certificate TypeCertificate of Analysis Date16/12/2022 Catalog Number252570024,252571000,252578000
Lot NumberA0233000 Certificate TypeCertificate of Analysis Date12/12/2022 Catalog Number252570024,252571000,252578000
Lot NumberA0375300 Certificate TypeCertificate of Analysis Date13/01/2021 Catalog Number252570024,252571000,252578000
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Ethylmagnesium chloride, 2.7M (25 wt.%) solution in THF
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
  • Substance which\, in contact with water\, emits flammable gases Category 1
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H351-Suspected of causing cancer.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
  • H260-In contact with water releases flammable gases which may ignite spontaneously.
  • H225-Highly flammable liquid and vapour.
  • EUH014-Reacts violently with water.
  • EUH019-May form explosive peroxides.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P231+P232-Handle under inert gas. Protect from moisture.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ethylmagnesium chloride, 2.7M (25 wt.%) solution in THF, AcroSeal™, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.